Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rac1b Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09271MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-271-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-271-MI Supplier MilliporeSigma Supplier No. 09271
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Rac1b using this Anti-Rac1b Antibody validated for use in western blot, immunohistochemistry (paraffin) and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rac1b
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in storage buffer containing PBS with 0.07% sodium azide in 30% glycerol.
Gene Symbols RAC1; MGC111543; ORFNames=MIG5; PAK1; MIG5; TC-25; p21-Rac1
Host Species Rabbit
Immunogen Synthetic peptide consisting of the internal region of Rac1b that is not in the other Rac1 isoforms.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_061485.1
Target Species Human, Mouse
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.