Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2), Mouse, Clone: 3B10-2D9, Abnova™

Catalog No. 89001483 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-483 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-483 Supplier Abnova Corporation Supplier No. H00005880M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RAC2.

The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. [provided by RefSeq

Sequence: MQAIKCVVVGDGAVGKTCLLISYTTNAFPGEYIPTVFDNYSANVMVDSKPVNLGLWDTAGQEDYDRLRPLSYPQTDVFLICFSLVSPASYENVRAKWFPEVRHHCPSTPIILVGTKLDLRDDKDTIEKLKEKKLAPITYPQGLALAKEIDSVKYLECSALTQRGLKTVFDEAIRAVLCPQPTRQQKRACSLL

Specifications

Antigen ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B10-2D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant RAC2.
Formulation ascites with no preservative
Gene RAC2
Gene Accession No. BC001485
Gene Alias EN-7/Gx/HSPC022
Gene Symbols RAC2
Host Species Mouse
Immunogen RAC2 (AAH01485.1, 1 a.a. ∼ 192 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5880
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.