Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAD18 homolog (S. cerevisiae), Mouse, Clone: 3H7, Abnova™

Catalog No. 89014321 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-321 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-321 Supplier Abnova Corporation Supplier No. H00056852M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RAD18.

The protein encoded by this gene is highly similar to S. cerevisiae DNA damage repair protein Rad18. Yeast Rad18 functions through its interaction with Rad6, which is an ubiquitin-conjugating enzyme required for post-replication repair of damaged DNA. Similar to its yeast counterpart, this protein is able to interact with the human homolog of yeast Rad6 protein through a conserved ring-finger motif. Mutation of this motif results in defective replication of UV-damaged DNA and hypersensitivity to multiple mutagens. [provided by RefSeq

Sequence: EKEIDEIHSKYRKKHKSEFQLLVDQARKGYKKIAGMSQKTVTITKEDESTEKLSSVCMGQEDNMTSVTNHFSQSKLDSPEELEPDREEDSSSCIDIQEV

Specifications

Antigen RAD18 homolog (S. cerevisiae)
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), KnockDown, Western Blot
Classification Monoclonal
Clone 3H7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RAD18.
Formulation PBS with no preservative; pH 7.4
Gene RAD18
Gene Accession No. NM_020165
Gene Alias RNF73
Gene Symbols RAD18
Host Species Mouse
Immunogen RAD18 (NP_064550, 332 a.a. ∼ 430 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56852
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.