Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad50 Mouse anti-Human, Unlabeled, Clone: 13, BD

Catalog No. BDB611010 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611010 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611010 Supplier BD Biosciences Supplier No. 611010

Mouse Monoclonal Antibody

DNA double-strand breaks (DSBs) are generated during intrinsic eukaryotic DNA recombination events such as assembly of antigen receptor genes, meiotic and miotic recombination. DNA DSB repair proteins are also required to repair breaks induced by extrinsic factors such as ionizing radiation and mutagenic chemicals. Originally identified in S. cerevisiae, Rad50 is one of a group of genes, designated as the Rad52 epistasis group, whose products mediate DSB repair. Many of these genes, including Rad50, are conserved in humans and are thought to have a similar function to their S. cerevisiae counterparts. In yeast, a multiprotein complex of Rad50, MRE11, and XRS2 has been implicated in the nucleocytic processing of DSBs. In humans, Rad50 and MRE11 complex with up to three additional proteins (95kDa, 200kDa, and 350kDa). The 95kDa species is thought to be human XRS2, although a separate report has identified it as Nibrin, the product of the gene mutated in Nijmegen breakage syndrome. The Rad50-MRE11-p95 complex possess endonuclease and 3′ to 5′ exonuclease activity. Thus, human Rad50 functions in a multiprotein complex to mediate the repair of DSBs in the human genome.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rad50
Applications Western Blot
Classification Monoclonal
Clone 13
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human RAD50 aa. 672-786
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.