Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad50 Mouse anti-Human, Mouse, Clone: 13B3/2C6, MilliporeSigma™

Catalog No. 05525MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-525-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-525-MI Supplier MilliporeSigma Supplier No. 05525
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Rad50 Antibody, clone 13B3/2C6 is a high quality Mouse Monoclonal Antibody for the detection of Rad50 and has been validated in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rad50
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 13B3/2C6
Conjugate Unconjugated
Formulation 10mM PBS, pH 7.4
Gene Accession No. NM_005732
Gene Symbols RAD50, RAD50-2, hRad50, hRAD50, RAD50-PEN
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 1-425 of human Rad50
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.