Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Rad51 (Ab-1), Polyclonal

Catalog No. PC130100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PC130100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PC130100UL Supplier MilliporeSigma™ Supplier No. PC130100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Rad51 Clone: in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunoprecipitation

The bacterial RecA and the yeast RAD51 proteins are involved in homologous recombination and DNA repair. The human RAD51 protein is the homologue of these two proteins. While the modular structure of these proteins is conserved and the homology is very strong with the bacterial protein there are functional differences seen with RAD51 including an ATP hydrolysis rate 1/10 that seen with RecA. Although RAD51 is a nuclear protein and is largely absent from the cytoplasm and nucleoli, the protein lacks a nuclear localization signal and requires other proteins in a complex to be translocated into the nucleus. Treatment of cells with DNA synthesis inducing agents will cause an increase in the levels of RAD51 protein which then undergo a shift in distribution becoming focal in nature. This focal distribution may be blocked by inhibitors of transcription. RAD51 can be shown to interact directly with the chromosomes and during the process of meiosis, its distribution of RAD51 undergoes dynamic change. RAD51 has been shown to interact with a wide variety of different proteins that include the wild-type form of p53, BRCA2, and RAD52, which specifically modulates its activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rad51
Applications Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 100mM Tris-Glycine, pH 7.4
Host Species Rabbit
Immunogen recombinant, human Rad51
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽43kDa Rad51 protein in ovarian tissue.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.