Learn More
RAD51 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA14985
Description
PA1-4985 detects Rad51 from human, mouse, and chicken samples. PA1-4985 has been successfully used in Western blot, immunoprecipitation, immunocytochemistry and indirect immunofluorescence applications. By Western blot, this antibody detects a 37 kDa band representing Rad51 from Hela cells. GFP-tagging of Rad51 protein at either N- or C- terminus inactivates the function of Rad51 and therefore it cannot be used for the study of foci formation instead of the antibody. The PA1-4985 immunogen is full-length recombinant Rap51 protein.
RAD51 plays a critical role in homologous strand exchange, a key step in DNA repair through homologous recobination. It binds to single and double-stranded DNA and exhibits DNA-dependent ATPase activity. RAD51 catalyzes the recognition of homology and strand exchange between homologous DNA partners to form a joint molecule between a processed DNA break and the repair template. It binds to a single-stranded DNA in an ATP-dependent manner to form nucleoprotein filaments which are essential for the homology search and strand exchange. Mutations in the gene can results in breast cancer, mirror movements 2 and fanconi anemia.
Specifications
RAD51 | |
Polyclonal | |
Unconjugated | |
Rad51 | |
AV304093; BRCA1/BRCA2-containing complex, subunit 5; BRCC5; DNA repair protein RAD51; DNA repair protein RAD51 homolog 1; DNA repair protein RAD51 homolog A; DNA repair protein rhp51; FANCR; homolog to S.cerevisiae; HRAD51; HsRad51; HsT16930; MGC128559 protein; MRMV2; MUT5; rad51; RAD51 homolog; RAD51 homolog (RecA homolog); RAD51 homolog (RecA homolog, E. coli); RAD51 homolog A; RAD51 recombinase; RAD51 recombinase L homeolog; rad51.1; rad51.L; rad51-1; RAD51A; rad51-a; rad51-b; Reca; RecA family recombinase Rhp51; RecA, E. coli, homolog of; RecA-like protein; recombinase RAD51; recombination protein A; RGD1563603; rhp51; SPAC644.14c; wu:fb38e12; xrad51; XRad51.1; YER095W; zgc:77754 | |
Rabbit | |
50 μL | |
Primary | |
Chicken, Human, Mouse, X. laevis | |
Antibody | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Conc. Not Determined | |
whole serum with 0.09% sodium azide | |
P37383, Q06609, Q08297 | |
Rad51 | |
Purified full-length recombinant Rad51 protein. | |
RUO | |
19361, 396086, 5888 | |
-20°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.