Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rad51D Mouse anti-Human, Clone: 5B3, MilliporeSigma™

Catalog No. MAB3698MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3698MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3698MI Supplier MilliporeSigma Supplier No. MAB3698

Mouse Monoclonal Antibody

Detect Rad51D with Anti-Rad51D Antibody, clone 5B3 (Mouse Monoclonal Antibody), that has been shown to work in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rad51D
Applications Western Blot
Classification Monoclonal
Clone 5B3
Conjugate Unconjugated
Formulation Ascites fluid. Liquid. Contains no preservative.
Gene Accession No. NM_002878.2; NM_133629.1
Gene Symbols RAD51L3, R51H3, Trad, TRAD, HsTRAD, RAD51D
Host Species Mouse
Immunogen Recombinant human Rad51D
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.