Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rae-1, Alexa Fluor 700, Clone: 186107, R&D Systems™

Catalog No. FAB17582N
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB17582N 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB17582N Supplier R&D Systems Supplier No. FAB17582N

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen Rae-1
Applications Flow Cytometry
Classification Monoclonal
Clone 186107
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q9JI58
Gene Symbols Raet1a
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Rae-1 delta , Leu29-Ser227, Accession # Q9JI58
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135250
Test Specificity Detects mouse Rae-1. It recognizes Rae-1 alpha , beta , delta , gamma and epsilon .
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.