Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAFT1 Mouse, Unlabeled, Clone: 30, BD

Catalog No. BDB611132 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611132 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611132 Supplier BD Biosciences Supplier No. 611132

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Immunosuppressive agents such as cyclosporin A, FK506, and rapamycin are used routinely in clinical transplantation and the treatment of autoimmune disorders. Cyclosporin A and FK506 bind to cyclophilins and the FK-binding protein FKBP12, respectively. These complexes interact with, and inhibit the activity of, the calcineurin protein phosphatase, which is essential for T cell activation. Rapamycin also interacts with FKBP12, but this complex does not affect calcineurin. Instead, it inhibits signals required for G1 to S progression in yeast, in
IL-2-stimulated T lymphocytes, and other mammalian cell types. Rapamycin-FKBP12 binds to a protein designated Rapamycin And FKBP12 Target-1 (RAFT1) in rats and FKBP-Rapamycin-Associated Protein (FRAP) in humans. RAFT1 is the homolog of the yeast genes TOR1 and TOR2 which, when mutated, lead to rapamycin resistance. RAFT1 is a PI4-kinase that exhibits homology with other PI kinases such as PI3-kinase and PI4-Kα. RAFT1 phosphorylates p70 S6 kinase and 4E-BP1 which are regulators of translational initiation. However, rapamycin-FKBP1 does not inhibit PI4-kinase activity and, therefore, affects RAFT1 activity via alternate mechanisms.

Host Species: Mouse
Clone: 30
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat RAFT1 aa. 185-290
Formula Weight [Chemical]: 245kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen RAFT1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 30
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Rat RAFT1 aa. 185-290
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.