Learn More
RAG-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE440
Description
Detect RAG-1 using this rabbit polyclonal antibody, Anti-RAG-1 Antibody validated for use in western blotting, immunocytochemistry and immunoprecipitation.
Specifications
RAG-1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
RAG1; RNF74 | |
KLH-conjugated linear peptide corresponding to a region near the N-terminus of human RAG-1. | |
100 μg | |
NP_000439 | |
2-8°C for 12 months |