Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ral A Mouse, Unlabeled, Clone: 8, BD

Catalog No. BDB610222 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610222 150 μg
BDB610221 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610222 Supplier BD Biosciences Supplier No. 610222
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ral is a low molecular weight GTP-binding protein belonging to the Ras superfamily of GTP-binding proteins and shows 50% amino acid identity to Ras. Ral cDNA clones have been isolated from human placenta, human pheochromocytoma, simian β-lymphocyte, and marine ray electric lobe cDNA libraries. In humans, cDNA sequences for Ral A and RalB have been determined. The predicted amino acid sequences show 85% identity. Both proteins consist of 206 amino acids with a predicted molecular weight of 24kDa. However, Ral A isolated from human platelets shows an apparent molecular weight of 26-28kDa. Ral A mRNA levels in adult mouse tissues appear to be highest in testes, ovary, and brain, with lower levels found in the liver, spleen, kidney, thymus, heart, and salivary glands. Similar results have been observed in rat tissues.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ral A
Applications Western Blot
Classification Monoclonal
Clone 8
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.