Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RalB Mouse anti-Human, Mouse, Rat, Clone: 25, MilliporeSigma™

Catalog No. 04037MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-037-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-037-MI Supplier MilliporeSigma Supplier No. 04037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect RalB using this Anti-RalB Antibody, clone 25 validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RalB
Applications Western Blot
Classification Monoclonal
Clone 25
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NM_002881.2
Gene Symbols RALB
Host Species Mouse
Immunogen Synthetic peptide of the C-terminus of RalB
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.