Learn More
RALY, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human RALY peptide using ARM Technology.
Supplier: Abnova Corporation H00022913K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
In infectious mononucleosis, anti-EBNA-1 antibodies are produced which cross-react with multiple normal human proteins. The cross-reactivity is due to anti-gly/ala antibodies that cross-react with host proteins containing configurations like those in the EBNA-1 repeat. One such antigen is RALY which is a member of the heterogeneous nuclear ribonucleoprotein gene family. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
RALY | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
RALY | |
RALY | |
A synthetic peptide of human RALY | |
RUO | |
22913 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog (mouse)) | |
RALY | |
HNRPCL2, MGC117312, P542 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.