Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ran Mouse, Unlabeled, Clone: 20, BD

Catalog No. BDB610340 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610340 50 μg
BDB610341 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610340 Supplier BD Biosciences Supplier No. 610340
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ran is a highly conserved GTPase that is ubiquitously expressed. At steady-state, 80-90% of cellular Ran is located in the nucleus, with the remainder in the cytoplasm. The human TC4/Ran cDNA was originally isolated based on the predicted homology of its encoded protein product to Ras. Unlike other members of the Ras superfamily, Ran does not appear to contain signals for lipid modification, nor does it appear to show membrane localization. Ran has been implicated in a number of cellular processes such as the initiation of DNA replication, entry into and exit from mitosis and in nuclear RNA and protein transport through the nuclear pore complex. A number of Ran binding proteins have been identified. One of the smaller proteins identified is a 28kDa cytosolic molecule known as RanBP1. The GTP-bound state of Ran is stabilized through its binding to RanBP1.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ran
Applications Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.