Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ran Rabbit anti-Human, Xenopus, Polyclonal, MilliporeSigma™

Catalog No. 07517MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-517-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-517-MI Supplier MilliporeSigma Supplier No. 07517

Rabbit Polyclonal Antibody

Anti-Ran Antibody detects level of Ran and has been published and validated for use in immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ran
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS and 0.1% sodium azide.
Gene Symbols RAN; Gsp1; ARA24; OK/SW-cl.81; TC4; RANBP3
Host Species Rabbit
Immunogen His-tagged recombinant human Ran protein purified from E. coli.
Purification Method Antiserum
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006325
Target Species Human, Xenopus species
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.