Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANTES Mouse anti-Human, Alexa Fluor 647, Clone: 2D5, BD

Catalog No. BDB564753 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564753 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564753 Supplier BD Biosciences Supplier No. 564753

Mouse Monoclonal Antibody

The 2D5 monoclonal antibody specifically binds to human Regulated upon Activation, Normal T cells Expressed and Secreted (RANTES). RANTES belongs to the C-C family of chemokines and is also known as CCL5. RANTES is produced by activated macrophages, CD8+ T cells, platelets, fibroblasts, epithelial cells and some tumor cells. RANTES is a chemoattractant and response modifier for a variety of cells including memory T cells, NK cells, dendritic cells, monocytes, eosinophils, basophils, and mast cells. RANTES binds to and transduces intracellular signals through several cell surface chemokine receptors including CCR1, CCR3, CCR4, and CCR5. RANTES can reportedly inhibit the interaction between certain HIV viruses and CCR5 and thus suppress viral infection in vitro.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RANTES
Applications Flow Cytometry
Classification Monoclonal
Clone 2D5
Conjugate Alexa Fluor 647
Description CCL5; chemokine (C-C motif) ligand 5; EoCP; SCYA5; SIS-delta; SISd; TCP228
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human RANTES Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.