Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANTES Mouse anti-Human, BB515, Clone: 2D5, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564752

Catalog No. BDB564752


Add to cart

Description

Description

The 2D5 monoclonal antibody specifically binds to human Regulated upon Activation, Normal T cells Expressed and Secreted (RANTES). RANTES belongs to the C-C family of chemokines and is also known as CCL5. RANTES is produced by activated macrophages, CD8+ T cells, platelets, fibroblasts, epithelial cells and some tumor cells. RANTES is a chemoattractant and response modifier for a variety of cells including memory T cells, NK cells, dendritic cells, monocytes, eosinophils, basophils, and mast cells. RANTES binds to and transduces intracellular signals through several cell surface chemokine receptors including CCR1, CCR3, CCR4, and CCR5. RANTES can reportedly inhibit the interaction between certain HIV viruses and CCR5 and thus suppress viral infection in vitro.

The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (eg, 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.

Specifications

Specifications

RANTES
2D5
CCL5; chemokine (C-C motif) ligand 5; EoCP; SCYA5; SIS-delta; SISd; TCP228
Affinity purified
Recombinant Human RANTES Protein
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide
Murine
IgG1, κ
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit