Learn More
CCL5 (RANTES), PE, clone: 21445, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA523619
Description
This gene is one of several CC cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The cytokine encoded by this gene functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.
In direct ELISAs and Western blots, this antibody does not cross-react with other chemokines, including recombinant human CCL2, 3, 4 recombinant mouse (rm) CCL3, and rmCCL4.
Specifications
CCL5 (RANTES) | |
Monoclonal | |
PE | |
CCL5 | |
C-C motif chemokine 5, SIS-delta, T-cell specific protein p288, T-cell-specific protein RANTES, beta-chemokine RANTES, eoCP, eosinophil chemotactic cytokine, small inducible cytokine A5, t cell-specific protein P228, D17S136E, MGC17164, SCYA5, SISd, TCP228 | |
Mouse | |
Protein A/G | |
RUO | |
6352 | |
Store at 4°C, store in the dark, do not freeze | |
Liquid |
Flow Cytometry | |
21445 | |
Buffered saline solution with BSA and 0.1% sodium azide | |
P13501 | |
CCL5 | |
E. coli-derived recombinant human CCL5/RANTES | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.