Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANTES Mouse anti-Human, PE, Clone: 2D5, BD

Catalog No. BDB554732 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554732 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554732 Supplier BD Biosciences Supplier No. 554732

Mouse Monoclonal Antibody

The 2D5 antibody reacts with human Regulated upon Activation, Normal T cells Expressed and Secreted (RANTES). The immunogen used to generated this hybridoma was recombinant human RANTES protein. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 2D5
Isotype: IgG1 κ
Species Reactivity: Human
Immunogen: Recombinant human RANTES protein

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen RANTES
Applications Flow Cytometry
Classification Monoclonal
Clone 2D5
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant human RANTES protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.