Learn More
Rap1 Mouse, Unlabeled, Clone: 3, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 610196
Description
Rap1 is a member of the large Ras superfamily of low molecular weight GTP/GDP binding proteins. Like Ras, the Rap proteins cycle between a GDP-bound inactive form and a GTP-bound active form. Since Ras and Rap have the same amino acid sequence in their putativeeffector domain (aa. 32-40), it seems likely that they perform either similar or antagonistic functions. Rap1A and Rap1B are highly homologous proteins, differing in only 9 of their 184 amino acids. Overexpression of Rap1A (also known as Krev-1) causes reversion of the phenotype of a Ki-Ras-transformed cell line. In vitro, Rap1 can compete efficiently with p21ras for interaction with Ras-GAP. Though they appear to have similar activities, Rap1 and Ras differ in their cellular localization. Ras is found on the inner surface of the plasma membrane while Rap1 is associated with the Golgi.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| Rap1 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Chicken, Frog, Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| 3 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.