Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1 Mouse anti-Human, Mouse, Clone: 4C8/1, MilliporeSigma™

Catalog No. 05911MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-911-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-911-MI Supplier MilliporeSigma Supplier No. 05911

Mouse Monoclonal Antibody

Anti-RAP1 Antibody, clone 4C8/1 is a high quality Mouse Monoclonal Antibody for the detection of RAP1 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP1
Applications Western Blot
Classification Monoclonal
Clone 4C8/1
Conjugate Unconjugated
Gene Accession No. NM_018975.2
Gene Symbols TERF2IP, hRap1, DRIP5, RAP1
Host Species Mouse
Immunogen Human Rap1 Clone 4C8/1
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C for up to 24 months.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.