Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rap1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07916MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-916-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-916-MI Supplier MilliporeSigma Supplier No. 07916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Rap1 Antibody is validated for use in western blotting for the detection of Rap1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rap1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in 0.02M phosphate buffer, pH 7.6, 0.25M NaCl, and 0.1% sodium azide.
Gene Symbols RAP1GDS1; MGC118859; GDS1; RAP1; MGC118861
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide (KYDPTIEDSYRKQVEVDC) corresponding to amino acids 31-48 human Rap1a.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_021159.3
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.