Learn More
RAP1A, member of RAS oncogene family, Mouse, Clone: 3E1, Abnova™
Shop All Abnova Corporation ProductsDescription
The product of this gene belongs to the family of RAS-related proteins. These proteins share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between RAP proteins and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. The product of this gene counteracts the mitogenic function of RAS because it can interact with RAS GAPs and RAF in a competitive manner. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | RAP1A, member of RAS oncogene family |
| Applications | ELISA, In situ PLA, Western Blot |
| Classification | Monoclonal |
| Clone | 3E1 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant RAP1A. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RAP1A |
| Gene Accession No. | BC014086 |
| Gene Alias | KREV-1/KREV1/RAP1/SMGP21 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.