Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1 GTPase activating protein, Mouse, Clone: 2F7, Abnova™

Catalog No. 89000930 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-930 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-930 Supplier Abnova Corporation Supplier No. H00005909M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RAP1GA1.

Sequence: MIEKMQGSRMDEQRCSFPPPLKTEEDYIPYPSVHEVLGREGPFPLILLPQFGGYWIEGTNHEITSIPETEPLQSPTTKVKLECNPTARIYRKHFLGKEHFNYYSLDAALGHLVFSLKYDVIGDQEHLRLLLRTKCRTYHDVIPISCLTEFPNVVQMAKLVCEDVNVDRFYPVLYPKASRLIVTFDEHVISNNFKFGVIYQKLGQTSEEELFSTNEESPAFVEFLEFLGQKVKLQDFKGFRGGLDVTHGQTGTESVYCNFRNKEIMFHVSTKLPYTEGDAQQLQRKRHIGNDIVAVVFQDENTPLVPDMIASNFLHAYVVVQAEGGGPDGPLYKVSVTARDDVPFFGPPLPDPAVFRKGPEFQEFLLTKLINAEYACYKAEKFAKLEERTRAALLETLYEELHIHSQSMMGLGGDEDKMENGSGGGGFFESFKRVIRSRSQSMDAMGLSNKKPNTVSTSHSGSFAPNNPDLAKAAGISLIVPGKSPTRKKSGPFGSRRSSAIGIENIQEVQEKRESPPAGQKTPDSGHVSQEPKSENSSTQSSPEMPTTKNRAETAAQRAEALKDFSRSSSSASSFASVVEETEGVDGEDTGLESVSSSGTPHKRDSFIYSTWLEDSVSTTSGGSSPGPSRSPHPDAGKLGDPACPEIKIQLEASEQHMPQLGC

Specifications

Antigen RAP1 GTPase activating protein
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant RAP1GA1.
Formulation PBS with no preservative; pH 7.4
Gene RAP1GAP
Gene Accession No. BC054490
Gene Alias KIAA0474/RAP1GA1/Rap1GAP1/rap1GAPII
Gene Symbols RAP1GAP
Host Species Mouse
Immunogen RAP1GA1 (AAH54490.1, 1 a.a. ∼ 663 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5909
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.