Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAP1, GTP-GDP dissociation stimulator 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016896 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-896 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-896 Supplier Abnova Corporation Supplier No. H00005910B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RAP1GDS1 protein.

Sequence: MADNLSDTLKKLKITAVDKTEDSLEGCLDCLLQALAQNNTETSEKIQASGILQLFATLLTPQSSCKAKVANIIAEVAKNEFMRIPCVDAGLISPLVQLLNSKDQEVLLQTGRALGNICYDSHEGRSAVDQAGGAQIVIDHLRSLCSITDPANEKLLTVFCGMLMNYSNENDSLQAQLINMGVIPTLVKLLGIHCQNAALTEMCLVAFGNLAELESSKEQFASTNIAEELVKLFKKQIEHDKREMIFEVLAPLAENDAIKLQLVEAGLVECLLEIVQQKVDSDKEDDITELKTGSDLMVLLLLGDESMQKLFEGGKGSVFQRVLSWIPSNNHQLQLAGALAIANFARNDANCIHMVDNGIVEKLMDLLDRHVEDGNVTVQHAALSALRNLAIPVINKAKMLSAGVTEAVLKFLKSEMPPVQFKLLGTLRMLIDAQEAAEQLGKNVKLVERLVEWCEAKDHAGVMGESNRLLSALIRHSKSKDVIKTIVQSGGIKHLVTMATSEHVIMQNEALVALALIAALELGTAEKDLESAKLVQILHRLLADERSAPEIKYNSMVLICALMGSECLHKEVQDLAFLDVVSKLRSHENKSVAQQASLTEQRLTVES

Specifications

Antigen RAP1, GTP-GDP dissociation stimulator 1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RAP1GDS1 protein.
Formulation PBS with no preservative; pH 7.4
Gene RAP1GDS1
Gene Accession No. NM_021159.3
Gene Alias GDS1/MGC118859/MGC118861/SmgGDS
Gene Symbols RAP1GDS1
Host Species Mouse
Immunogen RAP1GDS1 (NP_066982.2, 1 a.a. ∼ 607 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5910
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.