Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAPGEF1, Mouse, Clone: 3D10, Abnova™

Catalog No. 89022382 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-382 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-382 Supplier Abnova Corporation Supplier No. H00002889M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant RAPGEF1.

This gene encodes a human guanine nucleotide exchange factor. It transduces signals from CRK by binding the SH3 domain of CRK, and activating several members of the Ras family of GTPases. This signaling cascade that may be involved in apoptosis, integrin-mediated signal transduction, and cell transformation. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some variants has not been determined. [provided by RefSeq

Sequence: MGNAIEKQKPLKRSHLYPWKQDSQRSHLSSFTMKLMDKFHSPKIKRTPSKKGKPAEVSVKIPEKPVNKEATDRFLPEGYPLPLDLEQQAVEFMSTSAVASRSQRQKNLSW

Specifications

Antigen RAPGEF1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant RAPGEF1.
Formulation PBS with no preservative; pH 7.4
Gene RAPGEF1
Gene Accession No. BC041710
Gene Alias C3G/DKFZp781P1719/GRF2
Gene Symbols RAPGEF1
Host Species Mouse
Immunogen RAPGEF1 (AAH41710, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2889
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.