Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAPGEF5, Mouse, Clone: 2E4, Abnova™

Catalog No. 89002759 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-759 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-759 Supplier Abnova Corporation Supplier No. H00009771M09
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant RAPGEF5.

Members of the RAS (see HRAS; MIM 190020) subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as RAPGEF5, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation (Rebhun et al., 2000 [PubMed 10934204]).[supplied by OMIM

Sequence: MGSSRLRVFDPHLERKDSAAALSDRELPLPTFDVPYFKYIDEEDEDDEWSSRSQSSTEDDSVDSLLSDRYVVVSGTPEKILEHLLNDLHLEEVQDKETETLLDDFLLTYTVFMTTDDLCQALLRHYSAKKYQGKEENSDVPRRKRKVLHLVSQWIALYKDWLPEDEHSKMFLKTIYRNVLDDVYEYPILEKELKEFQKILGMHRRHTVDEYSPQKKNKALFHQFSLKENWLQHRGTVTETEEIFCHVYITEHSYVSVKAKVSSIAQEILLCSQLGKRVQLVKKFIKIAAHCKAQRNLNSFFAIVMGLNTASVSRLSQTWEKIPGKFRKLFSELESLTDPSLNHKAYRDAFKKMKPPKIPFVPLLLKDVTFIHEGNKTFLDNLVNFEKLHMIADTVRTLRHCRTNQFGDLSPKEHQELKSYVNHLYVIDSQQALFELSHRIEPRV

Specifications

Antigen RAPGEF5,
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant RAPGEF5.
Formulation PBS with no preservative; pH 7.4
Gene RAPGEF5
Gene Accession No. BC039203
Gene Alias GFR/KIAA0277/MR-GEF/REPAC
Gene Symbols RAPGEF5
Host Species Mouse
Immunogen RAPGEF5 (AAH39203, 1 a.a. ∼ 444 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9771
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.