Learn More
MilliporeSigma™ anti-Raptor, Clone: 1H6.2, ™
Supplier: MilliporeSigma™ 051470/DEL
Description
Raptor is a subunit of mTORC1 which regulates cell growth and survival in response to nutrient and hormonal signals. mTORC1 is activated in response to growth factors or amino-acids. Within mTORC1, Raptor positively regulates mTORC1 activity and functions as a scaffold for recruiting mTORC1 substrates.
Host Species: Mouse
Clone: 1H6.2
Isotype: IgG1, κ
Cross Reactivity: Human, Mouse
Immunogen: Recombinant protein corresponding to human Raptor.
Immunocytochemistry, Immunoprecipitation, Western Blotting
Specifications
| Raptor | |
| 1H6.2 | |
| Mouse | |
| 100 μg | |
| Human, Mouse | |
| IgG1 κ |
| Monoclonal | |
| Unconjugated | |
| Recombinant protein corresponding to human Raptor. | |
| Primary | |
| Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.