Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Raptor, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007983 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-983 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-983 Supplier Abnova Corporation Supplier No. H00057521B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RPTOR protein.

This gene encodes a component of a signaling pathway that regulates cell growth in response to nutrient and insulin levels. The encoded protein forms a stoichiometric complex with the mTOR kinase, and also associates with eukaryotic initiation factor 4E-binding protein-1 and ribosomal protein S6 kinase. The protein positively regulates the downstream effector ribosomal protein S6 kinase, and negatively regulates the mTOR kinase. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MESEMLQSPLLGLGEEDEADLTDWNLPLAFMKKRHCEKIEGSKSLAQSWRMKDRMKTVSVALVLCLNVGVDPPDVVKTTPCARLECWIDPLSMGPQKALETIGANLQKQYENWQPRARYKQSLDPTVDEVKKLCTSLRRNAKEERVLFHYNGHGVPRPTVNGEVWVFNKNYTQYIPLSIYDLQTWMGSPSIFVYDCSNAGLIVKSFKQFALQREQELEVAAINPNHPLAQMPLPPSMKNCIQLAACEATELLPMIPDLPADLFTSCLTTPIKIALRWFCMQKCVSLVPGVTLDLIEKIPGRLNDRRTPLGELNWIFTAITDTIAWNVLPRDLFQKLFRQDLLVASLFRNFLLAERIMRSYNCTPVSSPRLPPTYMHAMW

Specifications

Antigen Raptor
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RPTOR protein.
Formulation No additive
Gene RPTOR
Gene Accession No. BC064515
Gene Alias KOG1/Mip1
Gene Symbols RPTOR
Host Species Mouse
Immunogen RPTOR (AAH64515.1, 1 a.a. ∼ 379 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 57521
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.