Learn More
Description
Specifications
Specifications
| Antigen | Raptor |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified rabbit polyclonal IgG in 0.02M PBS, 0.25M sodium chloride containing 0.1% sodium azide and 50% glycerol. |
| Gene Symbols | KIAA1303; Raptor |
| Host Species | Rabbit |
| Immunogen | KLH conjugated peptide encompassing the N-terminal region of human Raptor. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.