Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAR alpha Mouse anti-Human, Rat, Clone: 763, MilliporeSigma™

Catalog No. MAB5346MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5346MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5346MI Supplier MilliporeSigma Supplier No. MAB5346
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-RAR alpha Antibody, clone 763 is a Mouse Monoclonal Antibody for detection of Retinoic Acid Receptor also known as RAR alpha and has been validated in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAR alpha
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 763
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 10mM HEPES (pH 7.5), 150mM NaCl, 100μg/mL BSA, 50% glycerol.
Gene Accession No. NM_001033603.1; NM_000964.2; NM_001024809.2
Gene Symbols RARA, RAR-alpha, NR1B1, RAR
Host Species Mouse
Immunogen Peptide from the N-terminus of human Retinoic Acid Receptor, alpha isotype.
Quantity 50 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.