Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-RAR beta, Clone: EPR2017, Rabbit Monoclonal,

Catalog No. MABE474MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE474MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE474MI Supplier MilliporeSigma™ Supplier No. MABE474

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

Antigen RAR beta
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone EPR2017
Conjugate Unconjugated
Formulation Rabbit Monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Host Species Rabbit
Immunogen Synthetic peptide corresponding to human RAR beta.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) RARB, HAP, NR1B2
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.