Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Ras (31-43), Polyclonal

Catalog No. 55357150UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
55-357-150UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 55-357-150UG Supplier MilliporeSigma™ Supplier No. 55357150UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Ras (31-43) Clone: in Bovine, Human, Mouse, Rat samples, and it is validated for Immuno Blotting

Ras proteins bind GDP and GTP with high affinity. This region of Ras is important in the binding of Ras to its effector, Raf1. This sequence (residues 31-43) is nearly 100% conserved between the different Ras encoded proteins (H-, K-, and N-Ras) and through phylogeny from slime mold to man.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ras (31-43)
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 50% glycerol, pH 7.2.
Host Species Rabbit
Immunogen a synthetic peptide [EYDPTIEDSYRKQ(C)] corresponding to amino acids 31-43 in the effector binding Loop (L2) of human H-, K-, and N-Ras, conjugated to KLH
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes all forms of wild type Ras, including the mutant G12V Ras protein.
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.