Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ras Mouse anti-Human, Mouse, Rat, Clone: 9A11.2, MilliporeSigma™

Catalog No. 051072MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-107-2MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-107-2MI Supplier MilliporeSigma Supplier No. 051072
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Ras Antibody, (K-, H-, N-), clone 9A11.2 is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Ras.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ras
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 9A11.2
Conjugate Unconjugated
Formulation Protein G purified mouse monoclonal in storage buffer containing 0.1M Tris-Glycine (pH 7.4), 15mM NaCl, and 0.05% NaN3.
Gene Accession No. NM_005343
Gene Symbols K-Ras, Ki-Ras, K-Ras2, Kras-2, p21B, KRAS, RASK2, HRAS, HA_RAS, N-RAS, H-RAS, NRAS, NRAS1, ALPS4
Host Species Mouse
Immunogen Full length recombinant GST-tagged human H-Ras.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.