Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ras-GAP Mouse, Unlabeled, Clone: 13, BD

Catalog No. BDB610040 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610040 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB610040 Supplier BD Biosciences Supplier No. 610040
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ras-GAP, a protein of approximately 120kDa, is thought to serve as a down-regulator of p21ras by stimulating its otherwise weak intrinsic GTPase activity. Both growth factor receptor and oncogene-encoded tyrosine kinases require functional membrane-associ

Host Species: Mouse
Clone: 13
Isotype: IgG1
Species Reactivity: Canine
Immunogen: Human Ras-GAP aa.381 to 390
Formula Weight [Chemical]: 120kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ras-GAP
Applications Western Blot
Classification Monoclonal
Clone 13
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Ras-GAP aa.381 to 390
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.