Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASA1, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002913 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-913 Supplier Abnova Corporation Supplier No. H00005921D01
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human RASA1 protein.

The protein encoded by this gene is located in the cytoplasm and is part of the GAP1 family of GTPase-activating proteins. The gene product stimulates the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. Mutations leading to changes in the binding sites of either protein are associated with basal cell carcinomas. Alternative splicing results in two isoforms where the shorter isoform, lacking the N-terminal hydrophobic region but retaining the same activity, appears to be abundantly expressed in placental but not adult tissues. [provided by RefSeq

Sequence: MKGWYHGKLDRTIAEERLRQAGKSGSYLIRESDRRPGSFVLSFLSQMNVVNHFRIIAMCGDYYIGGRRFSSLSDLIGYYSHVSCLLKGEKLLYPVAPPEPVEDRRRVRAILPYTKVPDTDEISFLKGDMFIVHNELEDGWMWVTNLRTDEQGLIVEDLVEEVGREEDPHEGKIWFHGKISKQEAYNLLMTVGQVCSFLVRPSDNTPGDYSLYFRTNENIQRFKICPTPNNQFMMGGRYYNSIGDIIDHYRKEQIVEGYYLKEPVPMQDQEQVLNDTVDGKEIYNTIRRKTKDAFYKNIVKKGYLLKKGKGKRWKNLYFILEGSDAQLIYFESEKRATKPKGLIDLSVCSVYVVHDSLFGRPNCFQIVVQHFSEEHYIFYFAGETPEQAEDWMKGLQAFCNLRKSSPGTSNKRLRQVSSLVLHIEEAHKLPVKHFTNPYCNIYLNSVQVAKTHAREGQNPVWSEEFVFDDLPPDINRFEITLSNKTKKSKDPDILFMRCQLSRLQKGHATDEWFLLSSHIPLKGIEPGSLRVRARYSMEKIMPEEEYSEFKELILQKELHVVYALSHVCGQDRTLLASILLRIFLHEKLESLLLCTLNDREISMEDEATTLFRATTLASTLMEQYMKATATQFVHHALKDSILKIMESKQSCELSPSKLEKNEDVNTNLTHLLNILSELVEKIFMASEILPPTLRYIYGCLQKSVQHKWPTNTTMRTRVVSGFVFLRLICPAILNPRMFNIISDSPSPIAARTLILVAKSVQNLANLVEFGAKEPYMEGVNPFIKSNKHRMIMFLDELGNVPELPDTTEHSRTDLSRDLAALHEICVAHSDELRTLSNERGAQQHVLKKLLAITELLQQKQNQYTKTNDVR

Specifications

Antigen RASA1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human RASA1 protein.
Formulation No additive
Gene RASA1
Gene Accession No. NM_022650
Gene Alias CM-AVM/CMAVM/DKFZp434N071/GAP/PKWS/RASA/RASGAP/p120GAP/p120RASGAP
Gene Symbols RASA1
Host Species Rabbit
Immunogen RASA1 (NP_072179.1, 1 a.a. ∼ 870 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5921
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.