Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASSF5, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017178 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-178 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-178 Supplier Abnova Corporation Supplier No. H00083593B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human RASSF5 protein.

This gene is a member of the Ras association domain family. It functions as a tumor suppressor, and is inactivated in a variety of cancers. The encoded protein localizes to centrosomes and microtubules, and associates with the GTP-activated forms of Ras, Rap1, and several other Ras-like small GTPases. The protein regulates lymphocyte adhesion and suppresses cell growth in response to activated Rap1 or Ras. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MTVDSSMSSGYCSLDEELEDCFFTAKTTFFRNAQSKHLSKNVCKPVEETQRPPTLQEIKQKIDSYNTREKNCLGMKLSEDGTYTGFIKVHLKLRRPVTVPAGIRPQSIYDAIKEVNLAATTDKRTSFYLPLDAIKQLHISSTTTVSEVIQGLLKKFMVVDNPQKFALFKRIHKDGQVLFQKLSIADRPLYLRLLAGPDTEVLSFVLKENETGEVEWDAFSIPELQNFLTILEKEEQDKIQQVQKKYDKFRQKLEEALRESQGKPG

Specifications

Antigen RASSF5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RASSF5 protein.
Formulation PBS with no preservative; pH 7.4
Gene RASSF5
Gene Accession No. NM_182665
Gene Alias MGC10823/MGC17344/Maxp1/NORE1/NORE1A/NORE1B/RAPL/RASSF3
Gene Symbols RASSF5
Host Species Mouse
Immunogen RASSF5 (NP_872606.1, 1 a.a. ∼ 265 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83593
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.