Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASSF8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220092
Description
RASSF8 Polyclonal specifically detects RASSF8 in Human samples. It is validated for Western Blot.Specifications
| RASSF8 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| C12orf2, carcinoma associated HOJ-1, Carcinoma-associated protein HOJ-1, chromosome 12 open reading frame 2, DKFZp434O0227, FLJ11542, HoJ-1, HOJ1, Ras association (RalGDS/AF-6) domain family (N-terminal) member 8, ras association domain-containing protein 8 | |
| Rabbit | |
| 48 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%), Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.88mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q8NHQ8 | |
| RASSF8 | |
| Recombinant protein encompassing a sequence within the center region of human RASSF8. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 11228 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction