Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2a Rat anti-Mouse, Unlabeled, Clone: R11-89, BD

Catalog No. BDB553446 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553446 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553446 Supplier BD Biosciences Supplier No. 553446
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The R11-89 antibody recognizes mouse IgG2a, with strong reactivity to the Igh-1[a] allotype and weaker reactivity to Igh-1[b]. It does not react with other Ig isotypes. Molecular genetic analyses suggest that the Igh-I[b] allele, which encodes IgG2a[b], i

Host Species: Rat
Clone: R11-89
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Pooled mouse Ig

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a
Applications ELISA
Classification Monoclonal
Clone R11-89
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Pooled mouse Ig
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.