Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APRIL/TNFSF13 Antibody (Sacha-2) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP197600
Description
APRIL/TNFSF13 Monoclonal specifically detects APRIL/TNFSF13 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
APRIL/TNFSF13 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
TNFSF13 | |
Recombinant human APRIL (aa 105-250) fused to human ACRP30headless (aa 16-108). | |
0.05 mg | |
Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology | |
8741 | |
Store at 4C. Do not freeze. | |
IgG2a |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Sacha-2 | |
Unconjugated | |
A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction