Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1 Antibody (MIH5), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143262PE
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human, Mouse samples. It is validated for Flow Cytometry, Product Image.Specifications
| PD-L1/B7-H1 | |
| Monoclonal | |
| PE | |
| Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG2a λ |
| Flow Cytometry | |
| MIH5 | |
| Flow Cytometry | |
| CD274 | |
| The immunogen for this antibody was B7H1. | |
| 0.1 mL | |
| Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
| 29126 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction