Learn More
CCR6 Antibody (18B9E6), Alexa Fluor™ 488, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP225224AF488
Catalog No. NBP225224X
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
CCR6 Monoclonal specifically detects CCR6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
CCR6
Monoclonal
Alexa Fluor 488
BN-1, CC-CKR-6, CCR-6, CD196 antigen, chemokine (C-C motif) receptor 6, CKR-L3G-protein coupled receptor 29, CKRL3LARC receptor, CMKBR6STRL22, DRY6, GPR29C-C CKR-6, GPRCY4DRY-6, lymphocyte, 22
Rat
Protein G purified
RUO
Primary
This product reacts with Mouse.
Store at 4C in the dark.
IgG2a κ
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
18B9E6
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CCR6
A recombinant protein consisting of the external domains of mouse CD196 (CCR6) was used as the immunogen for the antibody.
0.1 mL
Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
1235
Human, Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only