Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD97 Antibody (2Q42), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP222000UV
Description
CD97 Monoclonal specifically detects CD97 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD97 | |
Monoclonal | |
DyLight 350 | |
CD97 antigenseven-span transmembrane protein, CD97 molecule, Leukocyte antigen CD97, seven transmembrane helix receptor, seven-transmembrane, heterodimeric receptor associated with inflammation, TM7LN1 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
2Q42 | |
Flow Cytometry | |
ADGRE5 | |
Mouse recombinant CD97 | |
0.1 mL | |
GPCR, Immunology | |
976 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction