Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti- LIGHT/TNFSF14, Clone: Laury-1, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP197619
Description
LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
| LIGHT/TNFSF14 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:1000, Immunoprecipitation 1:200 | |
| TNFSF14 | |
| Recombinant human HVEM extracellular domain (aa 39-200). | |
| 0.05 mg | |
| Apoptosis | |
| 8740 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Flow Cytometry, Immunoprecipitation | |
| Laury-1 | |
| Unconjugated | |
| CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction