Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat T Lymphocyte Cocktail Rat, APC, FITC, PE, Clone: 1F4, OX-35, OX-8, BD

Catalog No. BDB558493 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558493 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558493 Supplier BD Biosciences Supplier No. 558493
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Cocktail Component Clone Isotype

APC anti-Rat CD3 1F4 mIgM

PE anti-Rat CD4 OX-35 mIgG2a

The Rat T Lymphocyte Cocktail is a three-color reagent cocktail designed to identify rat T lymphocytes by direct immunofluorescence staining with flow cytometric analysis. The OX-35 antibody reacts with the CD4 antigen on most thymocytes, a subpopulation of mature T lymphocytes (ie, MHC class II-restricted T cells, including most T helper cells), monocytes, macrophages, and some dendritic cells (1,2). The OX-8 antibody reacts with the hinge-like membrane-proximal domain of the 32-kDa alpha chain of the CD8 differentiation antigen (1,4). The CD8α and β chains (CD8a and CD8b, respectively) form a heterodimer on the surface of most thymocytes and a subpopuplation of mature T-lymphocytes (ie, MHC class I-restricted T cells, including most T suppressor/cytotoxic cells. The IF4 antibody reacts with the T-cell receptor-associated CD3 cell-surface antigen found on thymocytes and peripheral T lymphocytes (1,3).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat T Lymphocyte Cocktail
Applications Flow Cytometry
Classification Monoclonal
Clone 1F4, OX-35, OX-8
Conjugate FITC/APC/PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.