Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti- TIM-3, Clone: B8.2C12, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP236477
Description
TIM-3 Monoclonal specifically detects TIM-3 in Mouse samples. It is validated for Flow Cytometry.Specifications
TIM-3 | |
Monoclonal | |
1.0 mg/ml | |
Flow Cytometry 1ug/10^6cells | |
HAVCR2 | |
Recombinant mouse TIM-3 protein | |
0.1 mg | |
Cancer, Immunology, Innate Immunity, Neuroscience | |
84868 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Flow Cytometry | |
B8.2C12 | |
Unconjugated | |
CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction