Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TLR4 Antibody (MTS510), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP224865PCP
Description
TLR4 Monoclonal specifically detects TLR4 in Mouse samples. It is validated for Flow Cytometry, Flow (Cell Surface).Specifications
TLR4 | |
Monoclonal | |
PerCP | |
O00206 | |
TLR4 | |
This antibody was developed by immunizing rats with the Ba/F3 cell line expressing mouse TLR4 and MD-2 (Akashi et al, 2000). | |
0.1 mL | |
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Toll Like Receptors | |
7099 | |
Mouse | |
Purified |
Flow Cytometry, Blocking Assay | |
MTS510 | |
Flow Cytometry 1 μL/1 million cells, Flow (Cell Surface) | |
ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This antibody preferentially recognizes TLR4-MD-2 complex than of TLR4 alone. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction