Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti- TROY/TNFRSF19, Clone: Giu.Li, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP197623
Description
TROY/TNFRSF19 Monoclonal specifically detects TROY/TNFRSF19 in Human samples. It is validated for Flow Cytometry.Specifications
TROY/TNFRSF19 | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 1:10-1:1000 | |
TNFRSF19 | |
Recombinant human Troy cysteine-rich domain (aa 1-168). | |
0.025 mg | |
Apoptosis | |
55504 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Flow Cytometry | |
Giu.Li | |
Unconjugated | |
TAJ-alpha, TAJToxicity and JNK inducer, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction