Learn More
Rb (pS780) Mouse, PE, Clone: J146-35, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 558548
Description
The retinoblastoma gene product (Rb) is well known as a tumor suppressor and is either absent or mutated in many human tumors. Retrovirus-mediated gene transfer of the wild-type Rb gene into several Rb mutant neoplastic cell lines suppresses their tumorgenicity. Rb is a 110kDa nuclear phosphoprotein that undergoes differential phosphorylation during the cell cycle. During G1 phase, Rb is predominantly in a hypophosphorylated state. It becomes increasingly phosphorylated throughout the cell cycle until late mitosis, when substantial dephosphorylation occurs. Hypophosphorylated Rb interacts with a number of cellular proteins including the E2F transcription factor, several cyclins, RBP-1, RBP-2, c-Abl, c-myc, N-myc, and p46. Phosphorylation of Rb at various sites, by Cyclin-dependent protein kinases, inhibits the binding of Rb to these proteins. Rb is thought to mediate its effects, in part, via the repression of genes required for proliferation. For example, Rb is specifically recruited to promoters containing E2F sites and actively represses E2F mediated transcription. Rb also stimulates the activity of other transcription factors, although the mechanisms are less clearly defined. Thus, Rb appears to regulate transcription in its aim to control cell growth. The J146-35 monoclonal antibody recognizes Rb phosphorylated at serine 780 (pS780), which affects Rb binding to E2F. The orthologous phosphorylation sites in mouse and rat Rb are serines 773 and 751, respectively.
Intracellular Staining

Specifications
Rb (pS780) | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
J146-35 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Phosphorylated Human Rb Peptide | |
50 Tests | |
Cell Biology | |
Human, Mouse, Rat | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.